COM448658218 Combi-Blocks, Inc.

3,3-Diethoxy-propionic acid

  • MDL number MFCD06208054
  • Cas Number : 6191-97-5
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4821
Availability details for
Quantity
  • Availability
  • Add to Cart