COM448658240 Combi-Blocks, Inc.

2,4-Heptadien-6-one

  • MDL number MFCD00059238
  • Cas Number : 3916-64-1
  • Molecular Weight : 110.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-4845
Availability details for
Quantity
  • Availability
  • Add to Cart