COM448658383 Combi-Blocks, Inc.

4-Bromobenzoyl cyanide

  • MDL number MFCD00193627
  • Cas Number : 6048-21-1
  • Molecular Weight : 210.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5000
Availability details for
Quantity
  • Availability
  • Add to Cart