COM448658387 Combi-Blocks, Inc.

tert-Butyl 4-(4-chlorobenzoyl)piperazine-1-carboxylate

  • MDL number MFCD16620800
  • Cas Number : 897445-35-1
  • Molecular Weight : 324.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5004

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5004
Availability details for
Quantity
  • Availability
  • Add to Cart