COM448658417 Combi-Blocks, Inc.

5-(Piperazin-1-yl)pyrimidine hydrochloride

  • MDL number MFCD23141003
  • Cas Number : 634468-92-1
  • Molecular Weight : 200.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5035

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5035
Availability details for
Quantity
  • Availability
  • Add to Cart