COM448658514 Combi-Blocks, Inc.

3-(5-((2-Ethyl-1h-imidazol-1-yl)methyl)-1,2,4-oxadiazol-3-yl)benzaldehyde

  • MDL number MFCD18157700
  • Cas Number : 1258652-75-3
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5138

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5138
Availability details for
Quantity
  • Availability
  • Add to Cart