COM448658520 Combi-Blocks, Inc.

4-tert-Butoxybenzonitrile

  • MDL number MFCD03425273
  • Cas Number : 185259-36-3
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5145
Availability details for
Quantity
  • Availability
  • Add to Cart