COM448658538 Combi-Blocks, Inc.

5-Chlorothiazole

  • MDL number MFCD11053760
  • Cas Number : 4175-73-9
  • Molecular Weight : 119.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5163
Availability details for
Quantity
  • Availability
  • Add to Cart