COM448658561 Combi-Blocks, Inc.

7-Chloro-5-methyl-1h-pyrrolo[2,3-c]pyridine

  • MDL number MFCD11040277
  • Cas Number : 930790-43-5
  • Molecular Weight : 166.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5187

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5187
Availability details for
Quantity
  • Availability
  • Add to Cart