COM448658623 Combi-Blocks, Inc.

2-(2-Hydroxyethylamino)-n-(4-bromophenyl)acetamide

  • MDL number MFCD09037907
  • Cas Number : 1156583-05-9
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5257

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5257
Availability details for
Quantity
  • Availability
  • Add to Cart