COM448658647 Combi-Blocks, Inc.

Benzyl 2-naphthyl ether

  • MDL number MFCD00089092
  • Cas Number : 613-62-7
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5282

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5282
Availability details for
Quantity
  • Availability
  • Add to Cart