COM448658655 Combi-Blocks, Inc.

Quinoxaline-5-sulfonyl chloride

  • MDL number MFCD12827951
  • Cas Number : 844646-88-4
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5291

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5291
Availability details for
Quantity
  • Availability
  • Add to Cart