COM448658739 Combi-Blocks, Inc.

2,4-Dimethoxy-5-pyrimidinecarboxylic acid

  • MDL number MFCD03232921
  • Cas Number : 110821-07-3
  • Molecular Weight : 184.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5377
Availability details for
Quantity
  • Availability
  • Add to Cart