COM448658883 Combi-Blocks, Inc.

Methyl 4-fluoro-1H-pyrrole-2-carboxylate

  • MDL number MFCD11042594
  • Cas Number : 475561-89-8
  • Molecular Weight : 143.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5538

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5538
Availability details for
Quantity
  • Availability
  • Add to Cart