COM448658948 Combi-Blocks, Inc.

2,2,2-Trifluoroethoxybenzene

  • MDL number MFCD13193045
  • Cas Number : 17351-95-0
  • Molecular Weight : 176.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5609
Availability details for
Quantity
  • Availability
  • Add to Cart