COM448658970 Combi-Blocks, Inc.

(R)-4-Fluoro-indan-1-ylamine-HCl

  • MDL number MFCD16295030
  • Cas Number : 1637540-45-4
  • Molecular Weight : 187.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5631

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5631
Availability details for
Quantity
  • Availability
  • Add to Cart