COM448658983 Combi-Blocks, Inc.

Triphenylsulfonium 2,3,5,6-tetrafluoro-4-(methacryloyloxy)benzenesulfonate

  • MDL number MFCD11870892
  • Cas Number : 915090-37-8
  • Molecular Weight : 576.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5644
Availability details for
Quantity
  • Availability
  • Add to Cart