COM448659017 Combi-Blocks, Inc.

7-Chloro-3-methylbenzo[b]thiophene-2-carboxylic acid

  • MDL number MFCD11104814
  • Cas Number : 752135-41-4
  • Molecular Weight : 226.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5680
Availability details for
Quantity
  • Availability
  • Add to Cart