COM448659063 Combi-Blocks, Inc.

3-(2-Cyanophenyl)propanoic acid

  • MDL number MFCD00464857
  • Cas Number : 27916-43-4
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5730

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5730
Availability details for
Quantity
  • Availability
  • Add to Cart