COM448659065 Combi-Blocks, Inc.

6,8-Dichloro-2,7-naphthyridin-1(2h)-one hydrochloride

  • MDL number MFCD26405742
  • Cas Number : 1175559-45-1
  • Molecular Weight : 251.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5732
Availability details for
Quantity
  • Availability
  • Add to Cart