COM448659074 Combi-Blocks, Inc.

1-(2-Aminoethyl)-4-methoxycyclohexane

  • MDL number MFCD07772981
  • Cas Number : 1052223-70-7
  • Molecular Weight : 157.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5742
Availability details for
Quantity
  • Availability
  • Add to Cart