COM448659111 Combi-Blocks, Inc.

3,5-Difluoro-4-methylbenzonitrile

  • MDL number MFCD09835197
  • Cas Number : 170572-50-6
  • Molecular Weight : 153.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5779
Availability details for
Quantity
  • Availability
  • Add to Cart