COM448659195 Combi-Blocks, Inc.

Methyl 3-((furan-2-ylmethyl)amino)propanoate

  • MDL number MFCD01475841
  • Cas Number : 4063-31-4
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5867
Availability details for
Quantity
  • Availability
  • Add to Cart