COM448659238 Combi-Blocks, Inc.

5-Methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrazin-2-amine

  • MDL number MFCD17215187
  • Cas Number : 1227210-33-4
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5914
Availability details for
Quantity
  • Availability
  • Add to Cart