COM448659250 Combi-Blocks, Inc.

4-Chloro-3-isopropyl-1-methyl-1h-pyrazolo[3,4-b]quinoline

  • MDL number MFCD14585445
  • Cas Number : 884316-50-1
  • Molecular Weight : 259.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5926
Availability details for
Quantity
  • Availability
  • Add to Cart