COM448659292 Combi-Blocks, Inc.

Potassium 2-methyl-1-propenyltrifluoroborate

  • MDL number MFCD17677169
  • Cas Number : 1134643-87-0
  • Molecular Weight : 162.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5973

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-5973
Availability details for
Quantity
  • Availability
  • Add to Cart