COM448659338 Combi-Blocks, Inc.

Pyridoxamine dihydrochloride monohydrate

  • MDL number MFCD00150098
  • Cas Number : 524-36-7
  • Molecular Weight : 259.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6020

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6020
Availability details for
Quantity
  • Availability
  • Add to Cart