COM448659369 Combi-Blocks, Inc.

Acetyl isoniazid

  • MDL number MFCD00466330
  • Cas Number : 1078-38-2
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6052
Availability details for
Quantity
  • Availability
  • Add to Cart