COM448659391 Combi-Blocks, Inc.

(S)-1,4-Di-boc-piperazine-2-carboxylic acid methyl ester

  • MDL number MFCD18089802
  • Cas Number : 958635-19-3
  • Molecular Weight : 344.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6076
Availability details for
Quantity
  • Availability
  • Add to Cart