COM448659402 Combi-Blocks, Inc.

Potassium 2-methoxyethoxymethyltrifluoroborate

  • MDL number MFCD13195184
  • Cas Number : 910251-13-7
  • Molecular Weight : 196.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Freezer

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6087
Availability details for
Quantity
  • Availability
  • Add to Cart