COM448659403 Combi-Blocks, Inc.

2-(4-Bromo-2-hydroxyphenyl)acetic acid

  • MDL number MFCD18391507
  • Cas Number : 1261497-72-6
  • Molecular Weight : 231.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6088

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6088
Availability details for
Quantity
  • Availability
  • Add to Cart