COM448659450 Combi-Blocks, Inc.

4-Chloro-3h-imidazo[4,5-c]pyridine

  • MDL number MFCD13188423
  • Cas Number : 2770-01-6
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6137

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6137
Availability details for
Quantity
  • Availability
  • Add to Cart