COM448659464 Combi-Blocks, Inc.

1-Benzyl-2,5-dihydro-1h-pyrrole-3-carboxylic acid methyl ester

  • MDL number MFCD05662773
  • Cas Number : 101046-34-8
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6152
Availability details for
Quantity
  • Availability
  • Add to Cart