COM448659516 Combi-Blocks, Inc.

N-Cyclopropyl-1h-imidazole-1-carboxamide

  • MDL number MFCD11099699
  • Cas Number : 1033090-34-4
  • Molecular Weight : 151.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6208

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6208
Availability details for
Quantity
  • Availability
  • Add to Cart