COM448659521 Combi-Blocks, Inc.

1,1,3-Trifluoropropane

  • MDL number MFCD07784186
  • Cas Number : 24270-67-5
  • Molecular Weight : 98.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6213

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6213
Availability details for
Quantity
  • Availability
  • Add to Cart