COM448659590 Combi-Blocks, Inc.

2,6-Bis(difluoromethyl)pyran-4-one

  • MDL number MFCD08447319
  • Cas Number : 847947-32-4
  • Molecular Weight : 196.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6285
Availability details for
Quantity
  • Availability
  • Add to Cart