COM448659667 Combi-Blocks, Inc.

4-Bromo-3-(tributylstannyl)pyridine

  • MDL number MFCD09037862
  • Molecular Weight : 447.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6369

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6369
Availability details for
Quantity
  • Availability
  • Add to Cart