COM448659739 Combi-Blocks, Inc.

3-Methylnonane

  • MDL number MFCD00027308
  • Cas Number : 5911-04-6
  • Molecular Weight : 142.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6449
Availability details for
Quantity
  • Availability
  • Add to Cart