COM448659789 Combi-Blocks, Inc.

N2-Isobutyryl-2'-fluoro-2'-deoxyguanosine

  • MDL number MFCD15145246
  • Cas Number : 80681-25-0
  • Molecular Weight : 355.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6500
Availability details for
Quantity
  • Availability
  • Add to Cart