COM448659908 Combi-Blocks, Inc.

1-[3,5-Bis(trifluoromethyl)phenyl]propylamine

  • MDL number MFCD07784287
  • Cas Number : 685503-45-1
  • Molecular Weight : 271.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6630
Availability details for
Quantity
  • Availability
  • Add to Cart