COM448659911 Combi-Blocks, Inc.

(2R)-2-[(4-Ethyl-2,3-dioxopiperazinyl)carbonylamino]-2-phenylacetic acid

  • MDL number MFCD00799536
  • Cas Number : 63422-71-9
  • Molecular Weight : 319.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6633
Availability details for
Quantity
  • Availability
  • Add to Cart