COM448659937 Combi-Blocks, Inc.

3,3'-Di-tert-butyl-5,5'-dimethyldiphenoquinone

  • MDL number MFCD01310671
  • Cas Number : 2417-00-7
  • Molecular Weight : 324.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6663
Availability details for
Quantity
  • Availability
  • Add to Cart