COM448660081 Combi-Blocks, Inc.

Piperidin-3-yl-carbamic acid benzyl ester

  • MDL number MFCD04115317
  • Cas Number : 31648-54-1
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6818
Availability details for
Quantity
  • Availability
  • Add to Cart