COM448660088 Combi-Blocks, Inc.

5,5-Dimethyl-4-(pyrrolidin-1-yl)furan-2(5h)-one

  • MDL number MFCD11851387
  • Cas Number : 944591-46-2
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6825
Availability details for
Quantity
  • Availability
  • Add to Cart