COM448660121 Combi-Blocks, Inc.

Alpha-d(+)mannose 1-phosphate dipotassium salt

  • MDL number MFCD00069782
  • Cas Number : 71888-67-0
  • Molecular Weight : 336.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6861

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6861
Availability details for
Quantity
  • Availability
  • Add to Cart