COM448660222 Combi-Blocks, Inc.

1-(tert-Butyl)-3-methyl-1h-pyrazole-5-carbonyl chloride

  • MDL number MFCD00084915
  • Cas Number : 175277-10-8
  • Molecular Weight : 200.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6969

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6969
Availability details for
Quantity
  • Availability
  • Add to Cart