COM448660224 Combi-Blocks, Inc.

1-Benzoyl-piperidin-3-one

  • MDL number MFCD03790907
  • Cas Number : 67452-85-1
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6971
Availability details for
Quantity
  • Availability
  • Add to Cart