COM448660238 Combi-Blocks, Inc.

Ethyl 7-bromoquinoline-3-carboxylate

  • MDL number MFCD16657753
  • Cas Number : 1226762-74-8
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6986
Availability details for
Quantity
  • Availability
  • Add to Cart