COM448660296 Combi-Blocks, Inc.

3-Fluoro-5-trifluoromethylphenylsulphonyl chloride

  • MDL number MFCD06660226
  • Cas Number : 886499-99-6
  • Molecular Weight : 262.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7045
Availability details for
Quantity
  • Availability
  • Add to Cart