COM448660375 Combi-Blocks, Inc.

2-(Methylamino)pyrimidine-4-carboxaldehyde dimethyl acetal

  • MDL number MFCD10566061
  • Cas Number : 180869-38-9
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-7134
Availability details for
Quantity
  • Availability
  • Add to Cart